Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Cation exchange bulk packing material

Cation exchange bulk packing material

PRP-X200 bulk resin, 12 - 20 µm, 100 Å, 1 g
Codice catalogo 553-0009
Nome del fornitore:   HAMILTON BONADUZ
Cation exchange bulk packing material
Codice catalogo 553-0009
Nome del fornitore:  HAMILTON BONADUZ
Numero CAS:  
Cation exchange bulk packing material
Cation exchange bulk packing material

PRP-X200 bulk resin, 12 - 20 µm, 100 Å, 1 g

Numero CAS:  
Price
Richiedi prezzo
Log in per disponibilità
1 * 1 g
Product Details & Documents

Polymer bulk packing material for cation exchange applications.

Chromatographic capacity and efficiency tests are conducted on both bulk and packed column resins to ensure product integrity.

Specifiche

Specifiche

  • Dimensioni particelle:
    12 - 20 µm
  • Conf:
    1 g
  • Dimensioni pori:
    100 Å
  • Descrizione:
    PRP-X200 bulk resin

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.