Cation exchange bulk packing material
PRP-X200 bulk resin, 10 µm, 100 Å, 1 g
Codice catalogo 553-0008
Nome del fornitore: HAMILTON BONADUZ
Cation exchange bulk packing material
Codice catalogo 553-0008
Nome del fornitore: HAMILTON BONADUZ
Numero CAS:
Vai a
Product Details & Documents
Polymer bulk packing material for cation exchange applications.
Chromatographic capacity and efficiency tests are conducted on both bulk and packed column resins to ensure product integrity.
Specifiche
Specifiche
- Dimensioni particelle:10 µm
- Conf:1 g
- Dimensioni pori:100 Å
- Descrizione:PRP-X200 bulk resin



