Orders LinkContactUsLinkComponent
COD standards
COD standards
COD standard solution, 1300 mg/l, 500 mL
Catalog # 701-0046
Supplier:   REAGECON
COD standards
Catalog # 701-0046
Supplier:  REAGECON
CAS Number:  
COD standards
COD standard solution, 1300 mg/l, 500 mL
CAS Number:  
Price
SFr. 175.00
Log in for availability
1 * 500 mL
Product Details & Documents
Ideal for use as control standards to verify that correct analysis for COD has taken place.
Documents
Specifications
Specifications
  • Concentrazione:
    1300 mg/l
  • Conf:
    500 mL
  • Descrizione:
    COD standard solution