/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Il tuo profilo
Cart
Switzerland
Order Entry
Order Status
Il tuo profilo
Cart
Orders Link
ContactUsLinkComponent
Home
BUFFER SOLUTION PH 9.225 @ 20C NIST 1 * 500 mL
BUFFER SOLUTION PH 9.225 @ 20C NIST 1 * 500 mL
Print
BUFFER SOLUTION PH 9.225 @ 20C NIST 1 * 500 mL
Codice catalogo REGE109225
Nome del fornitore:
REAGECON
BUFFER SOLUTION PH 9.225 @ 20C NIST 1 * 500 mL
Codice catalogo REGE109225
Nome del fornitore:
REAGECON
Espandi l'immagine
Return to Top
BUFFER SOLUTION PH 9.225 @ 20C NIST 1 * 500 mL
Add to cart
Price
SFr. 43.10
Log in
per disponibilità
Unità di misura
1 * 500 mL
-
+
Add to cart
Return Policy
Product Details & Documents
Cleaning and Storage Solutions
#: 662-9223
Supplier: HAMILTON BONADUZ