Orders LinkContactUsLinkComponent
Stages
Stages
Stage micrometer 25 +50/10 mm - 25 lines with 1 mm intervals in x-direction - 50 lines with 1/10 mm intervals in y-direction
Codice catalogo 630-0424
Nome del fornitore:   ZEISS
Stages
Codice catalogo 630-0424
Nome del fornitore:  ZEISS
Stages
Stage micrometer 25 +50/10 mm - 25 lines with 1 mm intervals in x-direction - 50 lines with 1/10 mm intervals in y-direction
Price
329.65 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Specifiche
  • Descrizione:
    Stage micrometer 25 +50/10 mm - 25 lines with 1 mm intervals in x-direction - 50 lines with 1/10 mm intervals in y-direction
  • Seq:
    03