Accessories for on-line multi-parameter systems, IQ sensor net
Universal sensor extension assembly UA130 (incl. handle and set of seals), for mounting and protecting all 650, 690, 70X and 70X IQ series sensors, length without sensor: 1067 mm
Codice catalogo 662-0246
Nome del fornitore: WTW ONLINE
Accessories for on-line multi-parameter systems, IQ sensor net
Codice catalogo 662-0246
Nome del fornitore: WTW ONLINE
Vai a
Product Details & Documents
Specifiche
- Descrizione:Universal sensor extension assembly UA130 (incl. handle and set of seals), for mounting and protecting all 650, 690, 70X and 70X IQ series sensors, length without sensor: 1067 mm
- Seq:27