pH Combination electrodes
Sensolyt® PtA ORP single rod measuring chain**, Sensolyt® PtA (ORP), for installation into the SensoLyt® fitting, ±2000 mV, Acqua fortemente contaminata
Codice catalogo 708-0037
Nome del fornitore: WTW ONLINE
Vai a
Product Details & Documents
The SensoLyt® measuring chains with arming are easy to install and boast flexible measuring and application areas. Eletrodes without arming can be connected directly to a pH monitor of the EcoLine® or QuadroLine® series. Depending on the application, they can be installed into a flow vessel.
- Ideal for measurements in water and drinking water
- Also for high pressure and temperature conditions
- Easy to maintain by easy measuring chain replacement
Specifiche
- Per:Acqua fortemente contaminata
- Tipo:Sensolyt® PtA (ORP), for installation into the SensoLyt® fitting
- Descrizione:Sensolyt® PtA ORP single rod measuring chain**
- Seq:38
- pH:±2000 mV