pH Combination electrodes
Sensolyt® SE*, Sensolyt® SE, without arming, for the installation into flow vessels, 2 - 12, Acque reflue da moderatamente a fortemente inquinate
Catalog # 662-0280
Supplier: WTW ONLINE
Jump to:
Product Details & Documents
The SensoLyt® measuring chains with arming are easy to install and boast flexible measuring and application areas. Eletrodes without arming can be connected directly to a pH monitor of the EcoLine® or QuadroLine® series. Depending on the application, they can be installed into a flow vessel.
- Ideal for measurements in water and drinking water
- Also for high pressure and temperature conditions
- Easy to maintain by easy measuring chain replacement
Specifications
- Per:Acque reflue da moderatamente a fortemente inquinate
- Tipo:Sensolyt® SE, without arming, for the installation into flow vessels
- Descrizione:Sensolyt® SE*
- Seq:31
- pH:2 - 12