Plier decappers, WHEATON®
Pinza decapsulatrice per sigilli da 13 mm
Codice catalogo WHEA224372
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
These pliers decappers are constructed of steel with grey plastic coated handles. To remove a seal, simply secure the seal between the jaws, squeeze the handles and twist the seal.
Specifiche
- Descrizione:Pinza decapsulatrice per sigilli da 13 mm
- Seq:01