Palletank® Drum for Storage
Product Details & Documents
Palletank® for small volume storage are stainless steel containers designed for the reliable and robust storage of biopharmaceutical fluids such as high value drug substances contained in Flexsafe® 3D Bags. The Flexsafe® 3D Bags are manufactured according to a qualified design that precisely fits the Palletank®.
Applications:
Liquid storage and shipping
Media preparation
Buffer preparation
Drug substance purification
Drug product formulation
Cell harvest and downstream processing intermediates
The double-hinged front doors allow for bag replacement, installation, and manipulation while Palletank® are stacked up in the process area, thus enabling continuous processing without movement of the containers. As a result, a forklift is not routinely required in the manufacturing area.
Specifiche
- Ø est:918 mm (36,1")
- Capacità:200 L
- Descrizione:Drum with 1 bottom gate
- Materiale:HDPE
- Seq:2
- Cat. No.:SARTFBC125629
- Ø int.:559 mm (22")



