Orders LinkContactUsLinkComponent
TapeStation Platform Compliance Solution
TapeStation Platform Compliance Solution
Tapestation compliance solution for GLP/GMP environments
Codice catalogo AGLSSYS-TS-4200
New Product
Nome del fornitore:   AGILENT
TapeStation Platform Compliance Solution
Codice catalogo AGLSSYS-TS-4200
Nome del fornitore:  AGILENT
New Product
Numero CAS:  
TapeStation Platform Compliance Solution
Tapestation compliance solution for GLP/GMP environments
Numero CAS:  
Price
Richiedi prezzo
Log in per disponibilità
1 * 1 ST
Product Details & Documents
The Agilent tapestations compliance solution provides an integrated and reliable tool for quality control of nucleic acids. It supports the validation process from start to finish, enabling users to run the Agilent tapestation systems in regulated lab environments. The Agilent tapestation compliance solution comprises the 4150 and 4200 tapestation instrument, software, services, as well as application-specific consumables.
  • Reproducible results: System with prepackaged reagents and screentape devices, standardized protocols and optimized assays
    Objective information regarding sample concentration, size and integrity
    Compliance services: Software and hardware IQ and OQ can be purchased at any time. A certified engineer will test and verify the functionality of the hardware and software, thereby qualifying the system
    Design qualification: Functional and operational specifications are defined by the declaration of conformity for the tapestation instruments, screentape devices as well as the tapestation test tape, certificate of conformance for the reagents and the declaration of system validation for the Agilent tapestation software
Certifications: GLP/GMP compliant.
Specifiche
Specifiche
  • Descrizione:
    Tapestation compliance solution for GLP/GMP environments
  • Cat. No.:
    AGLSSYS-TS-4200
  • DNA:
    Yes