Kinetex Bulk Packing
XB-C18, 5 µm, 100 Å
Catalog # PHEC04A-4605
New Product
Supplier: PHENOMENEX
Jump to:
Product Details & Documents
Specifications
Specifications
- Dimensioni particelle:5 µm
- Dimensioni pori:100 Å
- Descrizione:XB-C18