Orders LinkContactUsLinkComponent
Kinetex Bulk Packing
XB-C18, 5 µm, 100 Å
Catalog # PHEC04A-4605
New Product
Supplier:   PHENOMENEX
Kinetex Bulk Packing
Catalog # PHEC04A-4605
Supplier:  PHENOMENEX
New Product
CAS Number:  
Kinetex Bulk Packing
Kinetex Bulk Packing
XB-C18, 5 µm, 100 Å
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
Specifications
  • Dimensioni particelle:
    5 µm
  • Dimensioni pori:
    100 Å
  • Descrizione:
    XB-C18