Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Bendamustine hydrochloride 99%

Bottle
Catalog # APOSOR1012316100MG
New Product
Supplier:   Apollo Scientific
Bendamustine hydrochloride 99%
Catalog # APOSOR1012316100MG
Supplier:  Apollo Scientific
New Product
CAS Number:  3543-75-7
Bendamustine hydrochloride 99%

Bottle

CAS Number:  3543-75-7
Price
SFr. 34.70
Log in for availability
1 * 100 mg
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Bendamustine HCl, 5-[Bis(2-chloroethyl)amino]-1-methylbenzimidazole-2-butyric acid hydrochloride, SDX-15 HCl, Cytostasane HCl, EP 311, 4-(5-(Bis(2-chloroethyl)amino)-1-methyl-1H-benzo[d]imidazol-2-yl)butanoic acid hydrochloride
  • CAS Number:3543-75-7
  • Formula:C₁₆H₂₂Cl₃N₃O₂
  • MDL Number:MFCD01658758
  • Molecular Weight:394,72 g/mol
Specifications

Specifications

  • Tipo di conf.:
    Bottle

Specifications