AC-Beta-ALA-OH 97%
Bottle
Codice catalogo APOSOR10121852.5KG
New Product
Nome del fornitore: Apollo Scientific
AC-Beta-ALA-OH 97%
Codice catalogo APOSOR10121852.5KG
Nome del fornitore: Apollo Scientific
New Product
Numero CAS: 3025-95-4
Vai a
Product Details & Documents
Usage: Organic building block/reagent
Sinonimi: N-Ac-β-Ala-OH, 3-Acetamidopropanoic acid, N-Acetyl-beta-alanine, N-Acetyl-b-alanine
- Numero CAS:3025-95-4
- Formula:C₅H₉NO₃
- Numero MDL:MFCD00037296
- Molecular Weight:131,13 g/mol
Specifiche
Specifiche
- Tipo di conf.:Bottle
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



