Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Bendamustine hydrochloride ≥95%

Bendamustine hydrochloride ≥95%

Bottle
Codice catalogo APOSBUP15045-1G
New Product
Nome del fornitore:   Apollo Scientific
Bendamustine hydrochloride ≥95%
Codice catalogo APOSBUP15045-1G
Nome del fornitore:  Apollo Scientific
New Product
Numero CAS:  3543-75-7
Bendamustine hydrochloride ≥95%

Bottle

Numero CAS:  3543-75-7
Price
752.00 SFr.
Log in per disponibilità
1 * 1 g
Product Details & Documents

Usage: Biochemical/life science applications.

Sinonimi:  Bendamustine HCl, 5-[Bis(2-chloroethyl)amino]-1-methylbenzimidazole-2-butyric acid hydrochloride, SDX-15 HCl, Cytostasane HCl, EP 311, 4-(5-(Bis(2-chloroethyl)amino)-1-methyl-1H-benzo[d]imidazol-2-yl)butanoic acid hydrochloride
  • Numero CAS:3543-75-7
  • Formula:C₁₆H₂₁Cl₂N₃O₂·HCl
  • Molecular Weight:394,72 g/mol
Specifiche

Specifiche

  • Tipo di conf.:
    Bottle

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.