Bendamustine hydrochloride ≥95%
Bottle
Catalog # APOSBUP15045-50MG
New Product
Supplier: Apollo Scientific
Bendamustine hydrochloride ≥95%
Catalog # APOSBUP15045-50MG
Supplier: Apollo Scientific
New Product
CAS Number: 3543-75-7
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications.
Synonyms: Bendamustine HCl, 5-[Bis(2-chloroethyl)amino]-1-methylbenzimidazole-2-butyric acid hydrochloride, SDX-15 HCl, Cytostasane HCl, EP 311, 4-(5-(Bis(2-chloroethyl)amino)-1-methyl-1H-benzo[d]imidazol-2-yl)butanoic acid hydrochloride
- CAS Number:3543-75-7
- Formula:C₁₆H₂₁Cl₂N₃O₂·HCl
- Molecular Weight:394,72 g/mol
Specifications
Specifications
- Tipo di conf.:Bottle



