Orders LinkContactUsLinkComponent
Cupferron
Bottle
  APOSOR2350-500G
New Product
Cupferron
  APOSOR2350-500G
New Product
 :  135-20-6
Cupferron
Bottle
 :  135-20-6
SFr. 91.80
1 * 500 g
 :  Ammonium 2-oxo-1-phenylhydrazinolate
  •  :135-20-6
  •  :C₆H₉N₃O₂
  •  :MFCD00078422
  •  :155.16 g/mol
  •  :154 °C (dec.)
  •  :Ambiente
  • Tipo di conf.:
    Bottle