Cupferron
Bottle
Catalog # APOSOR2350-5G
New Product
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: Ammonium 2-oxo-1-phenylhydrazinolate
- CAS Number:135-20-6
- Formula:C₆H₉N₃O₂
- MDL Number:MFCD00078422
- Molecular Weight:155.16 g/mol
- Melting Pt:154 °C (dec.)
- Storage Temperature:Ambiente
Specifications
Specifications
- Tipo di conf.:Bottle



