Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
(±)-α-Bromophenylacetic acid

(±)-α-Bromophenylacetic acid

Bottle
Catalog # APOSOR350067-5G
Corrosive
Irritant
New Product
Supplier:   Apollo Scientific
(±)-α-Bromophenylacetic acid
Catalog # APOSOR350067-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  4870-65-9
(±)-α-Bromophenylacetic acid

Bottle

CAS Number:  4870-65-9
Price
SFr. 29.50
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  α-Bromophenylacetic Acid, DL-α-Bromophenylacetic acid
  • CAS Number:4870-65-9
  • Formula:C₈H₇BrO₂
  • MDL Number:MFCD00004206
  • Molecular Weight:215.0440 g/mol
  • Melting Pt:82…83 °C
  • Storage Temperature:Ambiente
  • UN:3261
  • ADR:8,II
Specifications

Specifications

  • Tipo di conf.:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.