Orders LinkContactUsLinkComponent
(±)-α-Bromophenylacetic acid
Bottle
  APOSOR350067-500G
New Product
(±)-α-Bromophenylacetic acid
  APOSOR350067-500G
New Product
 :  4870-65-9
(±)-α-Bromophenylacetic acid
Bottle
 :  4870-65-9
1 170.00 SFr.
1 * 500 g
 :  α-Bromophenylacetic Acid, DL-α-Bromophenylacetic acid
  •  :4870-65-9
  •  :C₈H₇BrO₂
  •  :MFCD00004206
  •  :215.0440 g/mol
  •  :82…83 °C
  •  :Ambiente
  •  :3261
  •  :8,II
  • Tipo di conf.:
    Bottle