Reserpine 98%
Bottle
Catalog # APOSOR1030229-100M
New Product
Supplier: Apollo Scientific
Reserpine 98%
Catalog # APOSOR1030229-100M
Supplier: Apollo Scientific
New Product
CAS Number: 50-55-5
Jump to:
Product Details & Documents
Usage: Organic building block/reagent
- CAS Number:50-55-5
- Formula:C₃₃H₄₀N₂O₉
- MDL Number:MFCD00005091
- Molecular Weight:608,68 g/mol
- EINECS:200-047-9
Specifications
Specifications
- Tipo di conf.:Bottle



