Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Leflunomide 99%

Bottle
Codice catalogo APOSPC109034-100G
New Product
Nome del fornitore:   Apollo Scientific
Leflunomide 99%
Codice catalogo APOSPC109034-100G
Nome del fornitore:  Apollo Scientific
New Product
Numero CAS:  75706-12-6
Leflunomide 99%

Bottle

Numero CAS:  75706-12-6
Price
SFr. 2’030.00
Log in per disponibilità
1 * 100 g
Product Details & Documents

Usage: Fluorinated building block/ reagent.

Sinonimi:  5-Methyl-N-(4-(trifluoromethyl)phenyl)isoxazole-4-carboxamide, 5-Methyl-N-[4-(trifluoromethyl)phenyl]isoxazole-4-carboxamide
  • Numero CAS:75706-12-6
  • Formula:C₁₂H₉F₃N₂O₂
  • Molecular Weight:270,21 g/mol
Specifiche

Specifiche

  • Tipo di conf.:
    Bottle

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.