Leflunomide 99%
Bottle
Catalog # APOSPC109034-25G
New Product
Supplier: Apollo Scientific
Leflunomide 99%
Catalog # APOSPC109034-25G
Supplier: Apollo Scientific
New Product
CAS Number: 75706-12-6
Jump to:
Product Details & Documents
Usage: Fluorinated building block/ reagent.
Synonyms: 5-Methyl-N-(4-(trifluoromethyl)phenyl)isoxazole-4-carboxamide, 5-Methyl-N-[4-(trifluoromethyl)phenyl]isoxazole-4-carboxamide
- CAS Number:75706-12-6
- Formula:C₁₂H₉F₃N₂O₂
- Molecular Weight:270,21 g/mol
Specifications
Specifications
- Tipo di conf.:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



