Leflunomide 99%
Bottle
Codice catalogo APOSPC109034-5G
New Product
Nome del fornitore: Apollo Scientific
Leflunomide 99%
Codice catalogo APOSPC109034-5G
Nome del fornitore: Apollo Scientific
New Product
Numero CAS: 75706-12-6
Vai a
Product Details & Documents
Usage: Fluorinated building block/ reagent.
Sinonimi: 5-Methyl-N-(4-(trifluoromethyl)phenyl)isoxazole-4-carboxamide, 5-Methyl-N-[4-(trifluoromethyl)phenyl]isoxazole-4-carboxamide
- Numero CAS:75706-12-6
- Formula:C₁₂H₉F₃N₂O₂
- Molecular Weight:270,21 g/mol
Specifiche
Specifiche
- Tipo di conf.:Bottle
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



