Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Copper(II) methoxide

Copper(II) methoxide

Bottle
Catalog # APOSIN1564-5G
New Product
Supplier:   Apollo Scientific
Copper(II) methoxide
Catalog # APOSIN1564-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  76890-98-7
Copper(II) methoxide

Bottle

CAS Number:  76890-98-7
Price
SFr. 418.00
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Copper(II) dimethanolate
  • CAS Number:76890-98-7
  • Formula:Cu(CH₃O)₂
  • MDL Number:MFCD00015611
  • Molecular Weight:125.614 g/mol
  • Melting Pt:206 °C (dec.)
  • Storage Temperature:Ambiente
Specifications

Specifications

  • Tipo di conf.:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.