Copper(II) methoxide
Bottle
Catalog # APOSIN1564-1G
New Product
Supplier: Apollo Scientific
Copper(II) methoxide
Catalog # APOSIN1564-1G
Supplier: Apollo Scientific
New Product
CAS Number: 76890-98-7
Jump to:
Product Details & Documents
Synonyms: Copper(II) dimethanolate
- CAS Number:76890-98-7
- Formula:Cu(CH₃O)₂
- MDL Number:MFCD00015611
- Molecular Weight:125.614 g/mol
- Melting Pt:206 °C (dec.)
- Storage Temperature:Ambiente
Specifications
Specifications
- Tipo di conf.:Bottle