Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
3-Nitrobenzonitrile

3-Nitrobenzonitrile

Bottle
Catalog # APOSOR28671-500G
Irritant
New Product
Supplier:   Apollo Scientific
3-Nitrobenzonitrile
Catalog # APOSOR28671-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  619-24-9
3-Nitrobenzonitrile

Bottle

CAS Number:  619-24-9
Price
414.00 SFr.
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  m-Nitrobenzonitrile
  • CAS Number:619-24-9
  • Formula:C₇H₄N₂O₂
  • MDL Number:MFCD00007194
  • Molecular Weight:148.12 g/mol
  • Boiling Pt:165 °C/21mmHg
  • Melting Pt:114…117 °C
  • Storage Temperature:Ambiente
  • EINECS:210-587-7
  • UN:3439
  • ADR:6.1,II
Specifications

Specifications

  • Tipo di conf.:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.