Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Isonicotinic acid ≥98%

Bottle
  APOSBUP18513-5G
New Product
Isonicotinic acid ≥98%
  APOSBUP18513-5G
New Product
 :  55-22-1
Isonicotinic acid ≥98%

Bottle

 :  55-22-1
 
288.00 SFr.
   
1 * 5 g
 

Usage: Biochemical/life science applications

 :  Pyridine-4-carboxylic acid, 4-Pyridinecarboxylic acid, 4-Picolinic acid
  •  :55-22-1
  •  :C₆H₅NO₂
  •  :123,11 g/mol
 

 

  • Tipo di conf.:
    Bottle