Isonicotinic acid ≥98%
Bottle
Codice catalogo APOSBUP18513-500MG
New Product
Nome del fornitore: Apollo Scientific
Isonicotinic acid ≥98%
Codice catalogo APOSBUP18513-500MG
Nome del fornitore: Apollo Scientific
New Product
Numero CAS: 55-22-1
Vai a
Product Details & Documents
Usage: Biochemical/life science applications
Sinonimi: Pyridine-4-carboxylic acid, 4-Pyridinecarboxylic acid, 4-Picolinic acid
- Numero CAS:55-22-1
- Formula:C₆H₅NO₂
- Molecular Weight:123,11 g/mol
Specifiche
Specifiche
- Tipo di conf.:Bottle
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



