JK184 ≥98%
Bottle
Codice catalogo APOSBUP09972-50MG
New Product
Nome del fornitore: Apollo Scientific
JK184 ≥98%
Codice catalogo APOSBUP09972-50MG
Nome del fornitore: Apollo Scientific
New Product
Numero CAS: 315703-52-7
Vai a
Product Details & Documents
Usage: Biochemical/life science applications
Sinonimi: JK184, N-(4-Ethoxyphenyl)-4-(2-methylimidazo[1,2-a]pyridin-3-yl)thiazol-2-amine
- Numero CAS:315703-52-7
- Formula:C₁₉H₁₈N₄OS
- Molecular Weight:350,44 g/mol
Specifiche
Specifiche
- Tipo di conf.:Bottle
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



