Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
D-(-)-Mandelic acid ≥95%

D-(-)-Mandelic acid ≥95%

Bottle
Catalog # APOSBUP19814-5G
New Product
Supplier:   Apollo Scientific
D-(-)-Mandelic acid ≥95%
Catalog # APOSBUP19814-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  611-71-2
D-(-)-Mandelic acid ≥95%

Bottle

CAS Number:  611-71-2
Price
SFr. 83.20
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  D(-)-Mandelic acid, (R)-(-)-2-Hydroxy-2-phenylacetic acid, (R)-2-Hydroxy-2-phenylacetic acid, D(-)-2-Hydroxy-2-phenylacetic acid
  • CAS Number:611-71-2
  • Formula:C₈H₈O₃
  • Molecular Weight:152,15 g/mol
Specifications

Specifications

  • Tipo di conf.:
    Bottle

Specifications