Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
(-)-β-pinene ≥95%

(-)-β-pinene ≥95%

Bottle
Codice catalogo APOSBUP19725-5G
New Product
Nome del fornitore:   Apollo Scientific
(-)-β-pinene ≥95%
Codice catalogo APOSBUP19725-5G
Nome del fornitore:  Apollo Scientific
New Product
Numero CAS:  18172-67-3
(-)-β-pinene ≥95%

Bottle

Numero CAS:  18172-67-3
Price
SFr. 83.20
Log in per disponibilità
1 * 5 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Sinonimi:  (-)-Pin-2(10)-ene
  • Numero CAS:18172-67-3
  • Formula:C₁₀H₁₆
  • Molecular Weight:136,23 g/mol
Specifiche

Specifiche

  • Tipo di conf.:
    Bottle

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.