Orders LinkContactUsLinkComponent
Fluorescein ≥80%
Fluorescein ≥80%
Bottle
Catalog # APOSBUP19327-5G
New Product
Supplier:   Apollo Scientific
Fluorescein ≥80%
Catalog # APOSBUP19327-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  2321-07-5
Fluorescein ≥80%
Bottle
CAS Number:  2321-07-5
Price
SFr. 88.40
Log in for availability
1 * 5 g
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms:  3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid
  • CAS Number:2321-07-5
  • Formula:C₂₀H₁₂O₅
  • Molecular Weight:332,31 g/mol
Specifications
Specifications
  • Tipo di conf.:
    Bottle