Orders LinkContactUsLinkComponent
Fluorescein ≥80%
Bottle
  APOSBUP19327-1G
New Product
Fluorescein ≥80%
  APOSBUP19327-1G
New Product
 :  2321-07-5
Fluorescein ≥80%
Bottle
 :  2321-07-5
SFr. 64.10
1 * 1 g
Usage: Biochemical/ life science applications.
 :  3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid
  •  :2321-07-5
  •  :C₂₀H₁₂O₅
  •  :332,31 g/mol
  • Tipo di conf.:
    Bottle