AC-Beta-ALA-OH ≥98%
Bottle
Catalog # APOSBUP11881-200MG
New Product
Supplier: Apollo Scientific
AC-Beta-ALA-OH ≥98%
Catalog # APOSBUP11881-200MG
Supplier: Apollo Scientific
New Product
CAS Number: 3025-95-4
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications
Synonyms: N-Ac-β-Ala-OH, 3-Acetamidopropanoic acid, N-Acetyl-beta-alanine, N-Acetyl-b-alanine
- CAS Number:3025-95-4
- Formula:C₅H₉NO₃
- Molecular Weight:131,13 g/mol
Specifications
Specifications
- Tipo di conf.:Bottle



