Pimecrolimus 98%
Bottle
Catalog # APOSOR1026405-250M
New Product
Supplier: Apollo Scientific
Pimecrolimus 98%
Catalog # APOSOR1026405-250M
Supplier: Apollo Scientific
New Product
CAS Number: 137071-32-0
Jump to:
Product Details & Documents
Usage: Organic building block/reagent
Synonyms: (3S,4R,5S,8R,9E,12S,14S,15R,16S,18R,19R,26aS)-3-[(1E)-2-[(1R,3R,4S)-4-Chloro-3-methoxycyclohexyl]-1-methylethenyl]-8-ethyl-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-14,16-dimethoxy-4,10,12,18-tetramethyl-15,19-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclotricosine-1,7,20,21(4H,23H)-tetrone
- CAS Number:137071-32-0
- Formula:C₄₃H₆₈ClNO₁₁
- Molecular Weight:810,45 g/mol
Specifications
Specifications
- Tipo di conf.:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



