Pimecrolimus 98%
Bottle
Codice catalogo APOSOR1026405-100M
New Product
Nome del fornitore: Apollo Scientific
Pimecrolimus 98%
Codice catalogo APOSOR1026405-100M
Nome del fornitore: Apollo Scientific
New Product
Numero CAS: 137071-32-0
Vai a
Product Details & Documents
Usage: Organic building block/reagent
Sinonimi: (3S,4R,5S,8R,9E,12S,14S,15R,16S,18R,19R,26aS)-3-[(1E)-2-[(1R,3R,4S)-4-Chloro-3-methoxycyclohexyl]-1-methylethenyl]-8-ethyl-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-14,16-dimethoxy-4,10,12,18-tetramethyl-15,19-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclotricosine-1,7,20,21(4H,23H)-tetrone
- Numero CAS:137071-32-0
- Formula:C₄₃H₆₈ClNO₁₁
- Molecular Weight:810,45 g/mol
Specifiche
Specifiche
- Tipo di conf.:Bottle
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



