Orders LinkContactUsLinkComponent
Lanolin 98%
Lanolin 98%
Bottle
Catalog # APOSOR1024474-500G
Irritant
New Product
Supplier:   Apollo Scientific
Lanolin 98%
Catalog # APOSOR1024474-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  8006-54-0
Lanolin 98%
Bottle
CAS Number:  8006-54-0
Price
SFr. 69.30
Log in for availability
1 * 500 g
Product Details & Documents
Usage: Organic building block/reagent.
Synonyms:  Wool fat
  • CAS Number:8006-54-0
  • EINECS:232-348-6
Specifications
Specifications
  • Tipo di conf.:
    Bottle
Specifications