DL-Serina
Bottle
Catalog # APOSBIS2002-2.5KG
New Product
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: (±)-Serine, H-DL-Ser-OH, DL-2-Amino-3-hydroxypropionic acid
- CAS Number:302-84-1
- Formula:C₃H₇NO₃
- MDL Number:MFCD00064223
- Molecular Weight:105.09 g/mol
- Melting Pt:240 °C (dec.)
- Storage Temperature:Ambiente
Specifications
Specifications
- Tipo di conf.:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



