Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Acetohydroxamic acid

Acetohydroxamic acid

Bottle
Catalog # APOSBIA1030-2.5KG
Health
New Product
Supplier:   Apollo Scientific
Acetohydroxamic acid
Catalog # APOSBIA1030-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  546-88-3
Acetohydroxamic acid

Bottle

CAS Number:  546-88-3
Price
SFr. 1’440.00
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  N-Hydroxyacetamide
  • CAS Number:546-88-3
  • Formula:C₂H₅NO₂
  • MDL Number:MFCD00009994
  • Molecular Weight:75.0666 g/mol
  • Storage Temperature:Ambiente
Specifications

Specifications

  • Tipo di conf.:
    Bottle

Specifications