Acetohydroxamic acid
Bottle
Catalog # APOSBIA1030-100G
New Product
Supplier: Apollo Scientific
Acetohydroxamic acid
Catalog # APOSBIA1030-100G
Supplier: Apollo Scientific
New Product
CAS Number: 546-88-3
Jump to:
Product Details & Documents
Synonyms: N-Hydroxyacetamide
- CAS Number:546-88-3
- Formula:C₂H₅NO₂
- MDL Number:MFCD00009994
- Molecular Weight:75.0666 g/mol
- Storage Temperature:Ambiente
Specifications
Specifications
- Tipo di conf.:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



