phi29 Polymerase
Informazioni su questo articolo
Storage at −20 °C. DNA Polymerase 10 U/μl. Highly processive recombinant polymerase with exceptional strand displacement activity, which allows for highly efficient isothermal DNA amplification. The enzyme is capable of up to 70 thousands base insertions per binding event. The polymerase also possesses a 3'→5' proofreading exonuclease activity acting preferentially on ssDNA or ssRNA. Extremely high yields of amplified DNA can be obtained even from minute amounts of a template.
3 varianti
Product Details & Documents
Storage at −20 °C. DNA Polymerase 10 U/μl. Highly processive recombinant polymerase with exceptional strand displacement activity, which allows for highly efficient isothermal DNA amplification. The enzyme is capable of up to 70 thousands base insertions per binding event. The polymerase also possesses a 3'→5' proofreading exonuclease activity acting preferentially on ssDNA or ssRNA. Extremely high yields of amplified DNA can be obtained even from minute amounts of a template.
Specifications for Product Family
Specifiche
Specifiche
- Catalog No:
- APOSBIBL0054-1000U
- APOSBIBL0054-5000U
- APOSBIBL0054-250U
- Nome dell'enzima:
- phi29 Polymerase
- phi29 Polymerase
- phi29 Polymerase
Specifiche
Product Family Options
Informazioni sul prodotto
- DisponibilitàPrice
- Specifiche:
More
Specifications
Cat. No.APOSBIBL0054-1000UNome dell'enzimaphi29 Polymerase - Specifiche:
More
Specifications
Cat. No.APOSBIBL0054-5000UNome dell'enzimaphi29 Polymerase - Specifiche:
More
Specifications
Cat. No.APOSBIBL0054-250UNome dell'enzimaphi29 Polymerase