Orders LinkContactUsLinkComponent
Biozen Bulk Packing
Biozen Bulk Packing
Peptide XB-C18, 1,7 µm, 100 Å
Catalog # PHEC04A-4774
New Product
Supplier:   PHENOMENEX
Biozen Bulk Packing
Catalog # PHEC04A-4774
Supplier:  PHENOMENEX
New Product
CAS Number:  
Biozen Bulk Packing
Biozen Bulk Packing
Peptide XB-C18, 1,7 µm, 100 Å
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 pezzi
Product Details & Documents
Specifications
Specifications
  • Dimensioni particelle:
    1,7 µm
  • Dimensioni pori:
    100 Å
  • Descrizione:
    Peptide XB-C18