Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
EDTA

EDTA

EDTA
Codice catalogo EHERC13110800
Nome del fornitore:   EHRENSTORFER
EDTA
Codice catalogo EHERC13110800
Nome del fornitore:  EHRENSTORFER
Numero CAS:  
EDTA
EDTA

EDTA

Numero CAS:  
Price
SFr. 68.40
Log in per disponibilità
1 * 250 mg
Product Details & Documents
  • SMILES: OC(=O)CN(CCN(CC(=O)O)CC(=O)O)CC(=O)O
  • Inchi: InChI=1S/C10H16N2O8/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20)
  • IUPAC: 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
  • Packing group: III
Specifiche

Specifiche

  • Descrizione:
    EDTA
  • Peso molecolare:
    292,24
  • Cat. No.:
    EHERC13110800
  • Formula:
    C₁₀H₁₆N₂O₈
  • Condizioni di conservazione:
    20 °C
  • Descrizione classe di pericolo:
    GHS07, GHS08, GHS09
  • Segnale GHS:
    Attenzione
  • N. ONU:
    3077
  • Condizioni di spedizione:
    RT
  • Synonym(s):
    Glycine, N,N'-1,2-ethanediylbis[N-(carboxymethyl)-; Acetic acid, (ethylenedinitrilo)tetra- (8CI); N,N'-1,2-Ethanediylbis[N-(carboxymethyl)glycine]; 2,2',2'',2'''-(ethane-1,2-diylbis(azanetriyl))tetraacetic acid; 3,6-Diazaoctanedioic acid, 3,6-bis(carboxymethyl)-; Acetic acid, 2,2',2'',2'''-(1,2-ethanediyldinitrilo)tetrakis-; Acroma DH 700; Celon A; Celon ATH; Cheelox; Chelest 3A; Chelest A; Chemcolox 340; Clewat TAA; Clewat TTA; Complexon II; Dissolvine E; Dissolvine NA-X; Dissolvine Z; Dotite 4H; EDETA B; EDTA; EDTA (chelating agent); Edathamil; Edetic acid; Endrate; Ethylene-N,N'-biscarboxymethyl-N,N'-diglycine; Ethylenediamine-N,N,N',N'-tetraacetic acid; Ethylenediaminetetraacetate; Ethylenediaminetetraacetic acid; Ethylenedinitrilotetraacetic acid; Gluma Cleanser; Havidote; ICRF 185; Metaquest A; NSC 97243; NSC 97404; Nervanaid B acid; Nullapon B acid; Nullapon BF acid; Perma Kleer 50 acid; PrefGel; Quastal Special; Sequestrene AA; Sequestrene K 4; Sequestric acid; Sequestrol; Techrun DO; Titriplex; Titriplex II; Trilon BS; Trilon BW; Versene; WS; WS (chelating agent); YD 30; Zonon AO
  • CAS No.:
    60-00-4

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.