Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Bromacil

Bromacil

Bromacil
Catalog # EHERC10670000
Supplier:   EHRENSTORFER
Bromacil
Catalog # EHERC10670000
Supplier:  EHRENSTORFER
CAS Number:  
Bromacil
Bromacil

Bromacil

CAS Number:  
Price
SFr. 69.90
Log in for availability
1 * 0,25 g
Product Details & Documents
  • SMILES: CCC(C)N1C(=O)NC(=C(Br)C1=O)C
  • Inchi: InChI=1S/C9H13BrN2O2/c1-4-5(2)12-8(13)7(10)6(3)11-9(12)14/h5H,4H2,1-3H3,(H,11,14)
  • IUPAC: 5-bromo-3-butan-2-yl-6-methyl-1H-pyrimidine-2,4-dione
  • Packing group: III
Specifications

Specifications

  • Descrizione:
    Bromacil
  • Peso molecolare:
    261,12
  • Cat. No.:
    EHERC10670000
  • Formula:
    C₉H₁₃BrN₂O₂
  • Condizioni di conservazione:
    20 °C
  • Descrizione classe di pericolo:
    GHS07, GHS09
  • Segnale GHS:
    Attenzione
  • N. ONU:
    3077
  • Condizioni di spedizione:
    RT
  • Synonym(s):
    2,4(1H,3H)-Pyrimidinedione, 5-bromo-6-methyl-3-(1-methylpropyl)-; Uracil, 5-bromo-3-sec-butyl-6-methyl- (8CI); (±)-Bromacil; 3-sec-Butyl-5-bromo-6-methyluracil; 5-Bromo-3-sec-butyl-6-methyluracil; 5-Bromo-6-methyl-3-(1-methyl-n-propyl)uracil; 5-Bromo-6-methyl-3-(1-methylpropyl)uracil; 5-Bromo-6-methyl-3-sec-butyluracil; Bromacil; DuPont 976; Herbicide 976; Hyvar X; Hyvar X bromacil; Hyvarex; Uragan
  • CAS No.:
    314-40-9

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.