Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
L-Cysteic acid monohydrate ≥98%

L-Cysteic acid monohydrate ≥98%

Bottle
Catalog # APOSBUP19039-1ML
New Product
Supplier:   Apollo Scientific
L-Cysteic acid monohydrate ≥98%
Catalog # APOSBUP19039-1ML
Supplier:  Apollo Scientific
New Product
CAS Number:  23537-25-9
L-Cysteic acid monohydrate ≥98%

Bottle

CAS Number:  23537-25-9
Price
SFr. 45.10
Log in for availability
1 * 1 mL
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  2-Amino-3-sulphopropionic acid monohydrate
  • CAS Number:23537-25-9
  • Formula:C₃H₉NO₆S
  • Molecular Weight:187,17 g/mol
Specifications

Specifications

  • Tipo di conf.:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.