PHYTOSPHINGOSINE ≥98%
New Product
Nome del fornitore: Apollo Scientific
Vai a
Informazioni su questo articolo
Usage: Biochemical/ life science applications.
Sinonimi: (2S,3S,4R)-2-Aminooctadecane-1,3,4-triol, (2S,3S,4R)-2-Amino-1,3,4-octadecanetriol
- Numero CAS:554-62-1
- Formula:C₁₈H₃₉NO₃
- Molecular Weight:317,51 g/mol
3 varianti
Product Details & Documents
Usage: Biochemical/ life science applications.
Sinonimi: (2S,3S,4R)-2-Aminooctadecane-1,3,4-triol, (2S,3S,4R)-2-Amino-1,3,4-octadecanetriol
- Numero CAS:554-62-1
- Formula:C₁₈H₃₉NO₃
- Molecular Weight:317,51 g/mol
Product Family Options
Informazioni sul prodotto
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



