Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
(-)-β-pinene ≥95%

(-)-β-pinene ≥95%

New Product
Supplier:   Apollo Scientific
(-)-β-pinene ≥95%
Supplier:  Apollo Scientific
New Product
Certificates
(-)-β-pinene ≥95%

2 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  (-)-Pin-2(10)-ene
  • CAS Number:18172-67-3
  • Formula:C₁₀H₁₆
  • Molecular Weight:136,23 g/mol
Product Family Options

Product Information

  • Tipo di conf.
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - SFr. 83.20
    SFr. 83.20
    1 * 5 g
       SFr. 83.20

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - SFr. 149.00
    SFr. 149.00
    1 * 10 g
       SFr. 149.00

    Specifications:
    New Product
    New Product