Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
CHAPSO ≥98%

CHAPSO ≥98%

Bottle
Codice catalogo APOSBUP18607-500MG
New Product
Nome del fornitore:   Apollo Scientific
CHAPSO ≥98%
Codice catalogo APOSBUP18607-500MG
Nome del fornitore:  Apollo Scientific
New Product
Numero CAS:  82473-24-3
CHAPSO ≥98%

Bottle

Numero CAS:  82473-24-3
Price
91.80 SFr.
Log in per disponibilità
1 * 500 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Sinonimi:  (3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxypropanesulphonate), CHAPSO
  • Numero CAS:82473-24-3
  • Formula:C₃₂H₅₈N₂O₈S
  • Molecular Weight:630,88 g/mol
Specifiche

Specifiche

  • Tipo di conf.:
    Bottle

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.