Orders LinkContactUsLinkComponent
TES ≥98%
Bottle
Catalog # APOSBUP19670-1G
New Product
Supplier:   Apollo Scientific
TES ≥98%
Catalog # APOSBUP19670-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  7365-44-8
TES ≥98%
Bottle
CAS Number:  7365-44-8
Price
45.10 SFr.
Log in for availability
1 * 1 g
Product Details & Documents
Usage: Biochemical/life science applications.
Synonyms:  TES Buffer, N-Tris(hydroxymethyl)methyl-2-aminoethanesulphonic acid
  • CAS Number:7365-44-8
  • Formula:C₆H₁₅NO₆S
  • Molecular Weight:229,25 g/mol
Specifications
Specifications
  • Tipo di conf.:
    Bottle